Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318330.1 | NP_001305259.1 | 9 | 1041 | ||
NM_024948.3 | NP_079224.1 | 10 | 1093 | |||
XM_005252600.2 | XP_005252657.1 | 9 | 987 | |||
XM_005252602.2 | XP_005252659.1 | 8 | 935 | |||
XM_011519690.2 | XP_011517992.1 | 10 | 1070 | |||
XM_011519691.2 | XP_011517993.1 | 10 | 1070 | |||
XM_011519693.1 | XP_011517995.1 | 10 | 1079 | |||
XM_011519694.1 | XP_011517996.1 | 11 | 1134 | |||
XM_017016672.1 | XP_016872161.1 | 12 | 1226 | |||
XM_017016673.1 | XP_016872162.1 | 10 | 1096 | |||
GenBank mRNA | AB014761.1 | - | 10 | 859 | ||
AF063600.1 | - | 5 | 359 | |||
AF176916.1 | - | 9 | 1003 | |||
AK023459.1 | - | 10 | 1080 | |||
AL833873.1 | - | 10 | 1055 | |||
BC020605.1 | - | 9 | 1006 | |||
BC067799.1 | - | 10 | 1079 | 5278533 | ||
CR457315.1 | - | 10 | 859 |