Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024949.5 | NP_079225.5 | 7 | 965 | ||
XM_011532269.2 | XP_011530571.1 | 7 | 1598 | |||
GenBank mRNA | AK126057.1 | - | 6 | 630 | ||
BM046889.1 | - | 5 | 401 | |||
BX647378.1 | - | 5 | 556 |