Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
siRNA ID
HSS129305
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
5
683
NM_001007471.2
NP_001007472.2
5
1031
NM_020952.4
NP_066003.3
5
683
NM_024971.5
NP_079247.5
5
683
NM_206944.3
NP_996827.3
5
683
NM_206945.3
NP_996828.3
5
683
NM_206946.3
NP_996829.3
5
683
NM_206947.3
NP_996830.3
5
683
NM_206948.2
NP_996831.1
5
683
XM_011519035.2
XP_011517337.1
5
908
XM_011519036.2
XP_011517338.1
5
3833
XM_011519037.2
XP_011517339.1
5
3819
XM_011519038.2
XP_011517340.1
5
3816
XM_011519039.2
XP_011517341.1
5
3817
XM_011519040.2
XP_011517342.1
5
914
XM_011519041.2
XP_011517343.1
5
3816
XM_011519042.2
XP_011517344.1
5
914
XM_011519043.2
XP_011517345.1
5
947
XM_011519044.2
XP_011517346.1
5
3817
XM_011519045.2
XP_011517347.1
5
908
XM_011519046.2
XP_011517348.1
5
2611
XM_011519047.2
XP_011517349.1
5
2581
XM_017015141.1
XP_016870630.1
5
908
XM_017015142.1
XP_016870631.1
5
908
XM_017015143.1
XP_016870632.1
5
908
XM_017015144.1
XP_016870633.1
5
674
XM_017015145.1
XP_016870634.1
5
1352
XM_017015146.1
XP_016870635.1
5
750
XM_017015147.1
XP_016870636.1
5
2785
XM_017015148.1
XP_016870637.1
5
908
XM_017015149.1
XP_016870638.1
5
908
XM_017015150.1
XP_016870639.1
5
908
XM_017015151.1
XP_016870640.1
5
926
XM_017015152.1
XP_016870641.1
5
2586
XM_017015153.1
XP_016870642.1
5
918
XM_017015154.1
XP_016870643.1
5
1054
XM_017015155.1
XP_016870644.1
5
2583
XM_017015156.1
XP_016870645.1
1
106
XM_017015157.1
XP_016870646.1
5
2581
XM_017015158.1
XP_016870647.1
5
2579
XM_017015159.1
XP_016870648.1
5
674
XM_017015160.1
XP_016870649.1
5
764
XM_017015161.1
XP_016870650.1
5
908
GenBank mRNA
AB099661.1
-
2
328
AB099662.1
-
2
328
AB099663.1
-
2
328
AB099664.1
-
2
328
AB099665.1
-
2
328
AF325212.1
-
5
654
AF536748.1
-
2
328
AF536749.1
-
2
328
AF536750.1
-
2
328
AF536751.1
-
2
328
AF536752.1
-
2
328
AF536753.1
-
2
328
AJ505025.1
-
5
1031
AJ505026.1
-
5
1031
AK021788.1
-
5
674
AK225732.1
-
5
674
AK308682.1
-
5
859
AL136545.1
-
5
864
BC022454.2
-
4
613
BC067733.1
-
5
645
BC094699.1
-
5
677
BC121821.1
-
3
529
BC134414.1
-
3
532
BC142972.1
-
5
700
KF987075.1
-
5
739
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0