Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025003.3 | NP_079279.3 | 20 | 2846 | ||
XM_011538754.2 | XP_011537056.1 | 20 | 2899 | |||
XM_017019979.1 | XP_016875468.1 | 13 | 1915 | |||
GenBank mRNA | AF488804.1 | - | 20 | 2849 | ||
AJ515153.3 | - | 20 | 2840 | |||
AJ515154.2 | - | 20 | 2840 | |||
AK023228.1 | - | 3 | 344 |