Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320818.1 | NP_001307747.1 | 4 | 652 | ||
NM_001320819.1 | NP_001307748.1 | 5 | 846 | |||
NM_001320820.1 | NP_001307749.1 | 5 | 1043 | |||
NM_025147.4 | NP_079423.1 | 5 | 802 | |||
GenBank mRNA | AK023510.1 | - | 5 | 671 | ||
AK294093.1 | - | 5 | 1043 | |||
AK308977.1 | - | 5 | 640 | |||
BC022453.1 | - | 5 | 719 | 4796819 | ||
BU195968.1 | - | 4 | 586 |