Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201573.1 | NP_001188502.1 | 3 | 225 | ||
NM_025152.2 | NP_079428.2 | 5 | 461 | |||
NR_120408.1 | 5 | 461 | ||||
XM_011537182.2 | XP_011535484.1 | 6 | 435 | |||
XM_011537183.2 | XP_011535485.1 | 5 | 447 | |||
XM_017021664.1 | XP_016877153.1 | 5 | 447 | |||
XM_017021665.1 | XP_016877154.1 | 5 | 447 | |||
XM_017021666.1 | XP_016877155.1 | 5 | 447 | |||
GenBank mRNA | AK022722.1 | - | 5 | 374 | ||
AK295326.1 | - | 5 | 422 | |||
AK301447.1 | - | 3 | 225 | |||
AK316445.1 | - | 5 | 451 | |||
BC024919.1 | - | 5 | 387 | 4475196 | ||
BX248028.1 | - | 5 | 448 | |||
KJ534907.1 | - | 5 | 406 | |||
KJ534908.1 | - | 5 | 406 | |||
KJ534909.1 | - | 5 | 406 |