Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040451.2 | NP_001035541.1 | 13 | 1731 | ||
NM_001040452.2 | NP_001035542.1 | 14 | 1542 | |||
NM_025158.4 | NP_079434.3 | 14 | 1717 | |||
XM_005265993.4 | XP_005266050.1 | 11 | 1088 | |||
XM_006714921.3 | XP_006714984.2 | 13 | 1661 | |||
XM_006714922.3 | XP_006714985.1 | 11 | 1521 | |||
XM_017009891.1 | XP_016865380.1 | 13 | 1476 | |||
XM_017009892.1 | XP_016865381.1 | 12 | 1514 | |||
XM_017009893.1 | XP_016865382.1 | 11 | 1448 | |||
XM_017009894.1 | XP_016865383.1 | 15 | 1683 | |||
XM_017009895.1 | XP_016865384.1 | 14 | 1667 | |||
GenBank mRNA | AB209507.1 | - | 15 | 1587 | ||
AF361055.2 | - | 14 | 1690 | |||
AK025904.1 | - | 8 | 780 | |||
AK075021.1 | - | 13 | 1631 | |||
AK093848.1 | - | 6 | 1649 | |||
AK291926.1 | - | 13 | 1731 | |||
BC032571.1 | - | 14 | 1461 | 5243223 |