Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025194.2 | NP_079470.1 | 1 | 799 | ||
XM_017027324.1 | XP_016882813.1 | 1 | 27 | |||
GenBank mRNA | AJ290975.1 | - | 1 | 766 | ||
BC060788.1 | - | 1 | 799 | 30332196 | ||
D38169.1 | - | 1 | 530 |