Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195059.1 | NP_001181988.1 | 5 | 930 | ||
NM_025201.4 | NP_079477.2 | 6 | 1080 | |||
GenBank mRNA | AF318373.1 | - | 3 | 678 | ||
AF494534.1 | - | 6 | 1029 | |||
BC008744.2 | - | 5 | 891 | 3346334 |