Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143757.1 | NP_001137229.1 | 8, 9 | 1519 | ||
NM_001309474.1 | NP_001296403.1 | 7, 8 | 1276 | |||
NM_030626.2 | NP_085129.1 | 8, 9 | 1366 | |||
XM_006717983.2 | XP_006718046.1 | 8, 9 | 1679 | |||
XM_006717984.2 | XP_006718047.1 | 8, 9 | 1366 | |||
XM_011540208.1 | XP_011538510.1 | 8, 9 | 1449 | |||
XM_011540209.1 | XP_011538511.1 | 8, 9 | 1366 | |||
XM_011540210.2 | XP_011538512.1 | 7, 8 | 1274 | |||
XM_017016690.1 | XP_016872179.1 | 8, 9 | 1366 | |||
XM_017016694.1 | XP_016872183.1 | 7, 8 | 1253 | |||
GenBank mRNA | AB051461.1 | - | 8, 9 | 1276 | ||
AK091472.1 | - | 2, 3 | 1452 | |||
AK093250.1 | - | 8, 9 | 1132 | |||
AK097418.1 | - | 8, 9 | 1477 | |||
AK301422.1 | - | 7, 8 | 1043 | |||
BC058159.1 | - | 2, 3 | 253 |