Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSGA10 SIRNA Assays ›
Gene Symbol:
TSGA10
Gene Name:
testis specific 10
Gene Aliases:
CEP4L, CT79
Chromosome Location:
Chr.2: 98997261 - 99154962 on Build GRCh38
UniGene ID:
Hs.120267
Species:
Human
siRNA ID
HSS129654
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSGA10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025244.2
NP_079520.1
18
2279
NM_182911.3
NP_878915.2
17
2258
XM_005264023.1
XP_005264080.1
17
2233
XM_005264024.2
XP_005264081.1
19
2398
XM_005264025.1
XP_005264082.1
18
2369
XM_005264026.1
XP_005264083.1
19
2354
XM_005264027.1
XP_005264084.1
17
2189
XM_005264029.1
XP_005264086.1
17
2178
XM_006712781.2
XP_006712844.1
20
2432
XM_011511934.1
XP_011510236.1
17
2268
XM_011511935.2
XP_011510237.1
16
2042
XM_011511937.1
XP_011510239.1
17
2239
XM_011511939.1
XP_011510241.1
20
2527
XM_011511940.2
XP_011510242.1
19
2498
XM_011511941.1
XP_011510243.1
20
2571
XM_011511942.1
XP_011510244.1
20
2561
XM_011511943.2
XP_011510245.1
16
2009
XM_017005035.1
XP_016860524.1
17
2259
XM_017005039.1
XP_016860528.1
20
2421
XM_017005040.1
XP_016860529.1
19
2522
XM_017005041.1
XP_016860530.1
17
2380
XM_017005042.1
XP_016860531.1
19
2410
XM_017005043.1
XP_016860532.1
17
2243
XM_017005044.1
XP_016860533.1
19
2386
XM_017005045.1
XP_016860534.1
20
4573
XM_017005046.1
XP_016860535.1
19
2388
XM_017005047.1
XP_016860536.1
16
2296
XM_017005048.1
XP_016860537.1
16
2092
GenBank mRNA
AF254756.1
-
16
2254
AK313354.1
-
16
2015
AY014284.1
-
16
2107
BC028366.1
-
17
2259
4830604
BX647157.1
-
18
2472
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
spermatogenesis
cell projection assembly
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0