Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153448.3 | NP_703149.1 | 2 | 166 | ||
GenBank mRNA | AK097704.1 | - | 2 | 181 | ||
AY114148.1 | - | 2 | 140 | |||
BC042633.1 | - | 2 | 127 | 5170565 | ||
BC053599.1 | - | 2 | 155 | 5742902 |