Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITFG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305002.1 | NP_001291931.1 | 8 | 686 | ||
NM_030790.4 | NP_110417.2 | 8 | 963 | |||
GenBank mRNA | AF212247.1 | - | 8 | 871 | ||
AF503339.1 | - | 8 | 753 | |||
AK027596.1 | - | 8 | 754 | |||
AK299641.1 | - | 3 | 197 | |||
AK301908.1 | - | 8 | 638 | |||
AL833705.1 | - | 2 | 69 | |||
BC006321.2 | - | 8 | 736 | 4137934 | ||
BC024162.1 | - | 8 | 797 | 4046297 |