Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMAN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142292.1 | NP_001135764.1 | 7 | 772 | ||
NM_001322346.1 | NP_001309275.1 | 6 | 705 | |||
NM_001322347.1 | NP_001309276.1 | 6 | 669 | |||
NM_001322350.1 | NP_001309279.1 | 5 | 636 | |||
NM_001322351.1 | NP_001309280.1 | 4 | 553 | |||
NM_001322352.1 | NP_001309281.1 | 8 | 869 | |||
NM_001322354.1 | NP_001309283.1 | 6 | 719 | |||
NM_001322355.1 | NP_001309284.1 | 6 | 753 | |||
NM_001322356.1 | NP_001309285.1 | 5 | 672 | |||
NM_030805.3 | NP_110432.1 | 6 | 739 | |||
GenBank mRNA | AJ549957.1 | - | 6 | 703 | ||
AK075347.1 | - | 6 | 727 | |||
AK222828.1 | - | 6 | 717 | |||
AK295463.1 | - | 6 | 644 | |||
AK299740.1 | - | 6 | 677 | |||
AK301077.1 | - | 4 | 525 | |||
AK304515.1 | - | 5 | 607 | |||
AK308085.1 | - | 6 | 726 | |||
AK310100.1 | - | 6 | 756 | |||
AK312249.1 | - | 6 | 732 | |||
AK316331.1 | - | 5 | 644 | |||
AL136617.1 | - | 6 | 731 | |||
AY358929.1 | - | 6 | 710 | |||
BC000347.2 | - | 6 | 702 | |||
BC005822.2 | - | 6 | 702 | |||
BC005862.2 | - | 6 | 702 | |||
BC067265.1 | - | 6 | 709 | 6502212 | ||
BX648002.1 | - | 7 | 750 |