Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030815.2 | NP_110442.1 | 1, 2 | 212 | ||
GenBank mRNA | AK312021.1 | - | 1, 2 | 156 | ||
AY286301.1 | - | 1, 2 | 224 | |||
BC001856.2 | - | 1, 2 | 83 | |||
BC009334.2 | - | 1, 2 | 137 | 4126826 |