Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030816.4 | NP_110443.3 | 1 | 722 | ||
XM_005271234.3 | XP_005271291.1 | 1 | 965 | |||
XM_005271235.3 | XP_005271292.1 | 1 | 712 | |||
XM_006710929.3 | XP_006710992.1 | 1 | 735 | |||
XM_017002414.1 | XP_016857903.1 | 1 | 73 | |||
XM_017002415.1 | XP_016857904.1 | 1 | 73 | |||
GenBank mRNA | AL136717.1 | - | 1 | 723 | ||
BC016937.2 | - | 1 | 704 | |||
BC028840.1 | - | 1 | 711 | 4558748 | ||
BU622247.1 | - | 7 | 639 |
Gene Symbol: HHLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540547.2 | XP_011538849.1 | 1 | 170 |