Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135947.2 | NP_001129419.1 | 3, 4 | 261 | ||
NM_001265582.1 | NP_001252511.1 | 3 | 1624 | |||
NM_030914.3 | NP_112176.1 | 3, 4 | 261 | |||
NR_049743.1 | 3, 4 | 302 | ||||
GenBank mRNA | AK001880.1 | - | 3, 4 | 210 | ||
AK097029.1 | - | 3 | 1570 | |||
AK300877.1 | - | 3, 4 | 208 | |||
BC003581.1 | - | 3, 4 | 219 | 3546655 | ||
BI823604.1 | - | 3, 4 | 261 | |||
D83200.1 | - | 2, 3 | 167 | |||
DA691216.1 | - | 3, 4 | 267 |