Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NIPA2 SIRNA Assays ›
Gene Symbol:
NIPA2
Gene Name:
non imprinted in Prader-Willi/Angelman syndrome 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 22838641 - 22868384 on Build GRCh38
UniGene ID:
Hs.591003
Species:
Human
siRNA ID
HSS129977
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NIPA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008860.2
NP_001008860.1
7
1361
NM_001008892.2
NP_001008892.1
6
1225
NM_001008894.2
NP_001008894.1
5
1168
NM_001184888.1
NP_001171817.1
6
1316
NM_001184889.1
NP_001171818.1
10
1708
NM_030922.6
NP_112184.4
8
1483
XM_005272546.3
XP_005272603.1
8
1716
XM_005272547.4
XP_005272604.1
8
1500
XM_005272548.3
XP_005272605.1
7
1330
XM_005272550.3
XP_005272607.1
7
1168
XM_005272552.3
XP_005272609.1
6
1430
XM_005272553.4
XP_005272610.1
6
1234
XM_006720364.2
XP_006720427.1
9
1610
XM_006720365.2
XP_006720428.1
8
1455
XM_006720366.3
XP_006720429.1
7
1321
XM_006720367.1
XP_006720430.1
7
1659
XM_011543877.2
XP_011542179.1
9
1843
XM_011543878.2
XP_011542180.1
9
2010
XM_011543879.2
XP_011542181.1
9
1581
XM_011543880.2
XP_011542182.1
7
1594
XM_017022645.1
XP_016878134.1
9
1904
XM_017022646.1
XP_016878135.1
9
1671
XM_017022647.1
XP_016878136.1
9
1619
XM_017022648.1
XP_016878137.1
9
1680
XM_017022649.1
XP_016878138.1
9
1814
XM_017022650.1
XP_016878139.1
9
1777
XM_017022651.1
XP_016878140.1
10
2198
XM_017022652.1
XP_016878141.1
9
1590
XM_017022653.1
XP_016878142.1
10
1904
XM_017022654.1
XP_016878143.1
8
1428
XM_017022655.1
XP_016878144.1
7
1426
XM_017022656.1
XP_016878145.1
7
1436
XM_017022657.1
XP_016878146.1
8
1553
XM_017022658.1
XP_016878147.1
6
1274
XM_017022659.1
XP_016878148.1
8
1524
XM_017022660.1
XP_016878149.1
6
1537
XM_017022661.1
XP_016878150.1
6
1304
XM_017022662.1
XP_016878151.1
5
1373
XM_017022663.1
XP_016878152.1
5
1178
GenBank mRNA
AK096305.1
-
7
1342
AK300843.1
-
5
1111
AK313584.1
-
6
1300
AY732242.1
-
10
1629
BC000957.3
-
3
654
BC011775.2
-
6
1137
3640970
U90904.1
-
1
317
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
magnesium ion transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0