Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030960.2 | NP_112222.1 | 4 | 496 | ||
XM_011536160.2 | XP_011534462.1 | 4 | 450 | |||
XM_017011335.1 | XP_016866824.1 | 4 | 446 | |||
GenBank mRNA | AF203447.1 | - | 4 | 451 | ||
BC029488.1 | - | 4 | 496 | 5270534 |