Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031303.2 | NP_112593.2 | 11 | 1062 | ||
XM_005258357.4 | XP_005258414.1 | 14 | 1607 | |||
XM_005258358.4 | XP_005258415.1 | 14 | 1604 | |||
XM_005258361.3 | XP_005258418.1 | 10 | 864 | |||
XM_006722554.3 | XP_006722617.1 | 14 | 1918 | |||
XM_011526219.2 | XP_011524521.1 | 14 | 1607 | |||
XM_011526220.1 | XP_011524522.1 | 14 | 1607 | |||
XM_011526221.2 | XP_011524523.1 | 14 | 1607 | |||
XM_011526223.2 | XP_011524525.1 | 14 | 1607 | |||
XM_017026028.1 | XP_016881517.1 | 14 | 1607 | |||
XM_017026029.1 | XP_016881518.1 | 14 | 1918 | |||
XM_017026030.1 | XP_016881519.1 | 13 | 1558 | |||
XM_017026031.1 | XP_016881520.1 | 11 | 1043 | |||
XM_017026032.1 | XP_016881521.1 | 14 | 1648 | |||
GenBank mRNA | AK309134.1 | - | 4 | 698 | ||
AL512748.1 | - | 1 | 91 | |||
BC034999.2 | - | 11 | 1062 | 4829800 |