Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLVAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031310.2 | NP_112600.1 | 2, 3 | 532 | ||
GenBank mRNA | AF326591.1 | - | 2, 3 | 523 | ||
AF348827.1 | - | 2, 3 | 521 | |||
AK096030.1 | - | 2, 3 | 525 | |||
AK313837.1 | - | 2, 3 | 525 | |||
AL834363.1 | - | 2, 3 | 514 | |||
BC050365.1 | - | 2, 3 | 511 | 5761483 | ||
BC056414.1 | - | 2, 3 | 533 | 6190596 | ||
BX380375.2 | - | 2, 3 | 533 |