Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201366.1 | NP_001188295.1 | 4, 5 | 370 | ||
NM_031427.3 | NP_113615.2 | 3, 4 | 324 | |||
XM_017021679.1 | XP_016877168.1 | 4, 5 | 358 | |||
GenBank mRNA | AF542071.1 | - | 3, 4 | 200 | ||
AK315392.1 | - | 3, 4 | 219 | |||
BC005343.1 | - | 3, 4 | 200 | 3930213 |