Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF125106.1 | - | 6, 7 | 644 | ||
AF254133.1 | - | 6, 7 | 640 | |||
AK022317.1 | - | 6, 7 | 734 | |||
AK057848.1 | - | 7, 8 | 804 | |||
AK295471.1 | - | 6, 7 | 636 | |||
AK307475.1 | - | 6, 7 | 616 | |||
BC015547.1 | - | 5, 6 | 631 | 3845821 | ||
BC091495.1 | - | 6, 7 | 712 | 30396201 | ||
BI770572.1 | - | 6, 7 | 681 |