Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145313.2 | NP_001138785.1 | 5 | 560 | ||
NM_001287191.1 | NP_001274120.1 | 4 | 464 | |||
NM_001287192.1 | NP_001274121.1 | 4 | 464 | |||
NM_031919.4 | NP_114125.1 | 4 | 464 | |||
XM_005252254.2 | XP_005252311.1 | 5 | 793 | |||
XM_005252257.3 | XP_005252314.1 | 4 | 459 | |||
XM_005252260.3 | XP_005252317.1 | 4 | 449 | |||
XM_011519077.2 | XP_011517379.1 | 6 | 451 | |||
XM_011519078.2 | XP_011517380.2 | 5 | 458 | |||
XM_011519079.2 | XP_011517381.2 | 5 | 460 | |||
XM_017015182.1 | XP_016870671.1 | 5 | 793 | |||
XM_017015183.1 | XP_016870672.1 | 4 | 459 | |||
XM_017015185.1 | XP_016870674.1 | 5 | 466 | |||
XM_017015187.1 | XP_016870676.1 | 4 | 459 | |||
GenBank mRNA | AF088031.1 | - | 4 | 453 | ||
AF316830.1 | - | 4 | 327 | |||
AK296433.1 | - | 4 | 375 | |||
AK299350.1 | - | 5 | 492 | |||
AK299491.1 | - | 4 | 375 | |||
AK309817.1 | - | 5 | 801 | |||
BP219536.1 | - | 5 | 532 | |||
BP229804.1 | - | 4 | 464 | |||
BP365251.1 | - | 4 | 454 | |||
DN999402.1 | - | 4 | 307 |