Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193451.1 | NP_001180380.1 | 7 | 1294 | ||
NM_175861.3 | NP_787057.2 | 7 | 1385 | |||
XM_005253499.4 | XP_005253556.1 | 9 | 1546 | |||
XM_017020003.1 | XP_016875492.1 | 9 | 1547 | |||
XM_017020004.1 | XP_016875493.1 | 8 | 1339 | |||
XM_017020005.1 | XP_016875494.1 | 6 | 1152 | |||
XM_017020006.1 | XP_016875495.1 | 9 | 1550 | |||
XM_017020007.1 | XP_016875496.1 | 5 | 572 | |||
GenBank mRNA | AK055962.1 | - | 5 | 546 | ||
AK123619.1 | - | 3 | 976 | |||
AK127297.1 | - | 9 | 1356 | |||
BC042083.1 | - | 7 | 1385 | 5749757 | ||
GQ865601.1 | - | 7 | 1294 |