Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031935.2 | NP_114141.2 | 30 | 4752 | ||
XM_011510038.2 | XP_011508340.1 | 30 | 4769 | |||
XM_011510041.2 | XP_011508343.1 | 30 | 4777 | |||
XM_017002437.1 | XP_016857926.1 | 19 | 2702 | |||
GenBank mRNA | AF156100.1 | - | 30 | 4752 | ||
AK056557.1 | - | 13 | 1851 |