Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204240.1 | NP_001191169.1 | 9 | 1277 | ||
NM_031937.2 | NP_114143.1 | 9 | 1256 | |||
GenBank mRNA | AB449894.1 | - | 9 | 1171 | ||
AF331038.1 | - | 9 | 1195 | |||
AK074656.1 | - | 9 | 1222 | |||
AK093231.1 | - | 6 | 2391 | |||
AK127939.1 | - | 9 | 1212 | |||
AK131086.1 | - | 5 | 4821 | |||
AK309455.1 | - | 9 | 1022 | |||
BC022201.1 | - | 9 | 1205 | |||
BC136815.1 | - | 9 | 1212 | 9020806 | ||
BC142940.1 | - | 9 | 1228 | 8860306 | ||
BC150187.1 | - | 9 | 1176 | 40148357 | ||
BC150214.1 | - | 9 | 1181 | 40149007 | ||
BC171775.1 | - | 9 | 1212 | 9054429 | ||
CT841514.1 | - | 9 | 1177 |