Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031942.4 | NP_114148.3 | 8 | 1197 | ||
NM_145810.2 | NP_665809.1 | 7 | 960 | |||
XM_011511957.2 | XP_011510259.1 | 8 | 1738 | |||
GenBank mRNA | AK027628.1 | - | 8 | 1169 | ||
AK027642.1 | - | 7 | 941 | |||
AK075134.1 | - | 6 | 1823 | |||
AK223524.1 | - | 7 | 965 | |||
AK297097.1 | - | 6 | 795 | |||
AK297245.1 | - | 8 | 1057 | |||
AK300949.1 | - | 8 | 1026 | |||
AL833728.1 | - | 3 | 489 | |||
AY029179.1 | - | 7 | 973 | |||
BC015124.1 | - | 3 | 376 | |||
BC027966.1 | - | 7 | 926 | 5227112 |