Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317395.1 | NP_001304324.1 | 3 | 534 | ||
NM_001317399.1 | NP_001304328.1 | 3 | 534 | |||
NM_031954.4 | NP_114160.1 | 3 | 534 | |||
NR_133898.1 | 2 | 320 | ||||
GenBank mRNA | AF113208.1 | - | 3 | 279 | ||
AK027543.1 | - | 3 | 340 | |||
AK056912.1 | - | 3 | 1939 | |||
AK222548.1 | - | 3 | 290 | |||
AK309783.1 | - | 2 | 149 | |||
AY205299.1 | - | 3 | 340 | |||
AY597809.1 | - | 2 | 252 | |||
BC040062.1 | - | 3 | 287 | 5494016 | ||
BX333975.2 | - | 3 | 284 | |||
CX785497.1 | - | 2 | 63 | |||
DA440672.1 | - | 3 | 534 |