Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282546.1 | NP_001269475.1 | 5 | 755 | ||
NM_001282547.1 | NP_001269476.1 | 5 | 740 | |||
NM_032017.2 | NP_114406.1 | 6 | 977 | |||
GenBank mRNA | AK024504.1 | - | 4 | 539 | ||
AK075029.1 | - | 2 | 256 | |||
AK075048.1 | - | 5 | 737 | |||
AL834137.1 | - | 6 | 934 | |||
AY198395.1 | - | 5 | 710 | |||
BC007835.2 | - | 5 | 708 | 4109652 | ||
BC008344.1 | - | 5 | 716 | 3530483 |