Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032123.6 | NP_115499.5 | 11 | 1505 | ||
NM_199179.3 | NP_954648.3 | 10 | 1355 | |||
NM_199180.3 | NP_954649.3 | 11 | 1647 | |||
XM_011527362.1 | XP_011525664.1 | 12 | 1707 | |||
XM_011527363.1 | XP_011525665.1 | 12 | 1688 | |||
XM_011527365.1 | XP_011525667.1 | 11 | 1557 | |||
XM_011527366.1 | XP_011525668.1 | 10 | 1818 | |||
XM_017027348.1 | XP_016882837.1 | 11 | 2214 | |||
GenBank mRNA | AB593116.1 | - | 11 | 1611 | ||
AK307865.1 | - | 11 | 1968 | |||
AL136654.1 | - | 11 | 1631 | |||
AY305301.1 | - | 11 | 1449 | |||
AY305302.1 | - | 10 | 1299 | |||
AY358742.1 | - | 11 | 1633 | |||
BC064925.1 | - | 14 | 1570 | 6141879 |