Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029998.5 | NP_001025169.1 | 2 | 391 | ||
NM_001300842.2 | NP_001287771.1 | 2 | 391 | |||
NM_001317816.1 | NP_001304745.1 | 2 | 391 | |||
NM_001317817.1 | NP_001304746.1 | 2 | 391 | |||
NM_001317818.1 | NP_001304747.1 | 2 | 391 | |||
NM_032128.4 | NP_115504.1 | 2 | 391 | |||
NR_133924.1 | 2 | 391 | ||||
XM_011532311.2 | XP_011530613.1 | 2 | 391 | |||
XM_011532313.2 | XP_011530615.1 | 1 | 37 | |||
XM_017008689.1 | XP_016864178.1 | 2 | 391 | |||
XM_017008690.1 | XP_016864179.1 | 2 | 391 | |||
XM_017008691.1 | XP_016864180.1 | 2 | 391 | |||
XM_017008692.1 | XP_016864181.1 | 2 | 771 | |||
GenBank mRNA | AK291046.1 | - | 2 | 259 | ||
AL136728.1 | - | 2 | 319 | |||
AY346324.1 | - | 2 | 354 | |||
BC023288.1 | - | 2 | 342 | |||
BC063471.1 | - | 2 | 304 | |||
BC120970.1 | - | 2 | 250 | |||
BC120971.1 | - | 2 | 250 | |||
BC127626.1 | - | 2 | 250 | |||
BC127627.1 | - | 2 | 250 | |||
BC148252.1 | - | 2 | 250 | |||
BC150308.1 | - | 2 | 360 | 8860437 | ||
CR933647.1 | - | 2 | 385 | |||
CR936751.1 | - | 2 | 248 | |||
DA590625.1 | - | 2 | 392 | |||
DB205503.1 | - | 2 | 391 | |||
DQ122860.1 | - | 2 | 137 | |||
DQ122861.1 | - | 2 | 137 | |||
DQ871036.1 | - | 2 | 137 |