Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160184.1 | NP_001153656.1 | 4 | 431 | ||
NM_032129.2 | NP_115505.2 | 4 | 431 | |||
XM_006710944.3 | XP_006711007.2 | 4 | 1108 | |||
XM_011542248.2 | XP_011540550.2 | 4 | 1108 | |||
XM_017002474.1 | XP_016857963.1 | 4 | 1108 | |||
XM_017002475.1 | XP_016857964.1 | 4 | 1108 | |||
XM_017002476.1 | XP_016857965.1 | 4 | 1108 | |||
XM_017002477.1 | XP_016857966.1 | 4 | 1108 | |||
XM_017002478.1 | XP_016857967.1 | 4 | 1108 | |||
XM_017002479.1 | XP_016857968.1 | 4 | 1108 | |||
GenBank mRNA | AL136730.1 | - | 4 | 431 | ||
BC101386.2 | - | 4 | 426 | 40026400 | ||
BC101387.2 | - | 4 | 426 | 40026404 |