Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103511.1 | 6 | 905 | |||
XM_006713783.3 | XP_006713846.1 | 5 | 2309 | |||
XM_017007311.1 | XP_016862800.1 | 7 | 2488 | |||
XM_017007312.1 | XP_016862801.1 | 6 | 2543 | |||
XM_017007313.1 | XP_016862802.1 | 6 | 2544 | |||
GenBank mRNA | AK292958.1 | - | 5 | 569 | ||
AK310829.1 | - | 5 | 560 | |||
AL136815.1 | - | 4 | 336 | |||
BC024239.2 | - | 6 | 675 | 4826873 | ||
BM705017.1 | - | 6 | 526 |