Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032286.2 | NP_115662.2 | 1 | 115 | ||
GenBank mRNA | AY277599.1 | - | 1 | 72 | ||
AY453397.1 | - | 1 | 113 | |||
AY598325.1 | - | 1 | 72 | |||
BC003353.1 | - | 1 | 115 | 2899986 | ||
BI597082.1 | - | 1 | 104 |