Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GARNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286779.1 | NP_001273708.1 | 24 | 2492 | ||
NM_032293.4 | NP_115669.3 | 23 | 2629 | |||
NR_104591.1 | 23 | 2629 | ||||
XM_005252267.3 | XP_005252324.1 | 25 | 2484 | |||
XM_005252268.2 | XP_005252325.1 | 25 | 2546 | |||
XM_011519086.2 | XP_011517388.1 | 26 | 2556 | |||
XM_011519087.2 | XP_011517389.1 | 26 | 2539 | |||
XM_011519089.2 | XP_011517391.1 | 25 | 2442 | |||
XM_011519090.2 | XP_011517392.1 | 24 | 2325 | |||
XM_017015202.1 | XP_016870691.1 | 24 | 2522 | |||
GenBank mRNA | AK126704.1 | - | 2 | 3717 | ||
AK293732.1 | - | 23 | 2527 | |||
AK294970.1 | - | 23 | 2131 | |||
AK296246.1 | - | 24 | 2492 | |||
AL136573.1 | - | 23 | 2183 | |||
AL834149.1 | - | 7 | 761 | |||
BC034983.2 | - | 23 | 2239 | 4821143 |