Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318011.1 | NP_001304940.1 | 3 | 466 | ||
NM_032310.4 | NP_115686.3 | 3 | 466 | |||
NR_134464.1 | 3 | 477 | ||||
NR_134465.1 | 3 | 466 | ||||
NR_134466.1 | 3 | 477 | ||||
NR_134467.1 | 4 | 568 | ||||
NR_134468.1 | 3 | 477 | ||||
GenBank mRNA | AA804861.1 | - | 3 | 421 | ||
AK057716.1 | - | 3 | 372 | |||
AK091611.1 | - | 3 | 383 | |||
AK097977.1 | - | 1 | 769 | |||
AK128259.1 | - | 1 | 1504 | |||
BC004500.2 | - | 4 | 335 | 3833318 | ||
BC038856.1 | - | 3 | 359 | 5215640 | ||
BC091476.1 | - | 3 | 320 | |||
BM930683.1 | - | 3 | 311 | |||
BX354102.2 | - | 4 | 448 | |||
CD676329.1 | - | 3 | 333 |