Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZDHHC16 SIRNA Assays ›
Gene Symbol:
ZDHHC16
Gene Name:
zinc finger DHHC-type containing 16
Gene Aliases:
APH2, DHHC-16
Chromosome Location:
Chr.10: 97446131 - 97457370 on Build GRCh38
UniGene ID:
Hs.76662
Species:
Human
siRNA ID
HSS130872
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZDHHC16
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287803.1
NP_001274732.1
9
1200
NM_032327.3
NP_115703.2
8
1020
NM_198043.2
NP_932160.1
7
972
NM_198044.2
NP_932161.1
7
903
NM_198045.2
NP_932162.1
6
777
NM_198046.2
NP_932163.1
9
1200
NR_109895.1
6
577
XM_006718021.1
XP_006718084.1
8
1008
XM_006718022.1
XP_006718085.1
9
1111
XM_006718023.1
XP_006718086.1
7
960
XM_006718024.1
XP_006718087.1
8
1008
XM_006718025.1
XP_006718088.1
7
891
XM_006718026.1
XP_006718089.1
6
843
XM_017016766.1
XP_016872255.1
8
1063
XM_017016767.1
XP_016872256.1
7
960
XM_017016768.1
XP_016872257.1
8
994
XM_017016769.1
XP_016872258.1
6
843
XM_017016770.1
XP_016872259.1
7
778
XM_017016772.1
XP_016872261.1
6
730
XM_017016773.1
XP_016872262.1
9
1199
XM_017016774.1
XP_016872263.1
9
1199
GenBank mRNA
AB209272.1
-
6
2726
AF258563.1
-
7
901
AJ294945.1
-
9
1034
AK074427.1
-
8
974
AK297962.1
-
9
1156
AY359080.1
-
7
863
BC004535.2
-
6
698
3160750
BC008074.2
-
8
978
3029918
BC011749.2
-
8
966
3140177
BC012830.2
-
8
978
3029918
BC033157.1
-
7
830
4309312
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0