Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143853.1 | NP_001137325.1 | 2 | 107 | ||
NM_032345.2 | NP_115721.1 | 2 | 246 | |||
GenBank mRNA | AJ459406.1 | - | 2 | 94 | ||
AK096922.1 | - | 2 | 94 | |||
BC006135.1 | - | 2 | 246 | 3640407 | ||
BC014976.1 | - | 2 | 215 | 4854916 | ||
DB466388.1 | - | 2 | 107 |