Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRMS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032352.3 | NP_115728.2 | 1, 2 | 257 | ||
XM_005268128.1 | XP_005268185.1 | 1, 2 | 330 | |||
GenBank mRNA | AK310359.1 | - | 1, 2 | 191 | ||
AK315424.1 | - | 1, 2 | 188 | |||
AY827074.1 | - | 1, 2 | 243 | |||
BC006250.2 | - | 1, 2 | 237 | 3947694 | ||
BU507838.1 | - | 1, 2 | 301 |