Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256549.1 | NP_001243478.1 | 7 | 856 | ||
NM_001257101.1 | NP_001244030.1 | 7 | 749 | |||
NM_032373.4 | NP_115749.2 | 7 | 914 | |||
XM_006718030.1 | XP_006718093.1 | 6 | 845 | |||
XM_011540271.2 | XP_011538573.1 | 7 | 915 | |||
XM_011540272.2 | XP_011538574.1 | 7 | 946 | |||
XM_017016776.1 | XP_016872265.1 | 7 | 861 | |||
XM_017016777.1 | XP_016872266.1 | 7 | 860 | |||
XM_017016778.1 | XP_016872267.1 | 6 | 845 | |||
GenBank mRNA | AK303014.1 | - | 7 | 857 | ||
AL832003.2 | - | 5 | 353 | |||
AL832496.1 | - | 7 | 730 | |||
BC051845.1 | - | 7 | 724 | 5199133 |