Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032390.4 | NP_115766.3 | 1 | 133 | ||
GenBank mRNA | AB044971.1 | - | 1 | 110 | ||
AK291903.1 | - | 1 | 101 | |||
AK299814.1 | - | 1 | 83 | |||
AK311033.1 | - | 1 | 78 | |||
BC022990.1 | - | 1 | 89 | 4997441 | ||
BC024238.2 | - | 1 | 98 | 4797633 | ||
DA570244.1 | - | 1 | 133 |