Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEGF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256545.1 | NP_001243474.1 | 3, 4 | 536 | ||
NM_001308119.1 | NP_001295048.1 | 4, 5 | 587 | |||
NM_001308121.1 | NP_001295050.1 | 3, 4 | 536 | |||
NM_032446.2 | NP_115822.1 | 4, 5 | 587 | |||
XM_011543694.1 | XP_011541996.1 | 3, 4 | 299 | |||
XM_017009987.1 | XP_016865476.1 | 5, 6 | 832 | |||
GenBank mRNA | AB058676.1 | - | 4, 5 | 428 | ||
AK123568.1 | - | 5, 6 | 702 | |||
AK307919.1 | - | 3, 4 | 455 | |||
BC020198.1 | - | 4, 5 | 504 | |||
BC152478.1 | - | 4, 5 | 428 | 8862616 | ||
CR749437.1 | - | 3, 4 | 470 | |||
DA240513.1 | - | 3, 4 | 536 |