Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NTNG2 SIRNA Assays ›
Gene Symbol:
NTNG2
Gene Name:
netrin G2
Gene Aliases:
LHLL9381, Lmnt2, NTNG1, bA479K20.1
Chromosome Location:
Chr.9: 132161234 - 132244534 on Build GRCh38
UniGene ID:
Hs.163642
Species:
Human
siRNA ID
HSS131136
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NTNG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_032536.2
NP_115925.2
2, 3
992
XM_006717304.3
XP_006717367.1
4, 5
1013
XM_011519094.2
XP_011517396.1
4, 5
1013
XM_011519096.2
XP_011517398.1
4, 5
1410
XM_011519097.2
XP_011517399.1
3, 4
931
XM_011519098.2
XP_011517400.1
4, 5
963
XM_011519099.2
XP_011517401.1
3, 4
934
XM_011519100.2
XP_011517402.1
2, 3
863
XM_011519102.2
XP_011517404.1
3, 4
878
XM_011519103.2
XP_011517405.1
2, 3
770
XM_011519104.2
XP_011517406.1
2, 3
994
XM_011519105.2
XP_011517407.1
2, 3
860
XM_011519106.2
XP_011517408.1
4, 5
1013
XM_011519107.2
XP_011517409.1
4, 5
1013
XM_011519108.2
XP_011517410.1
4, 5
1013
XM_011519109.2
XP_011517411.1
4, 5
1013
XM_011519110.2
XP_011517412.1
4, 5
1013
XM_017015212.1
XP_016870701.1
5, 6
1117
XM_017015213.1
XP_016870702.1
3, 4
1320
XM_017015214.1
XP_016870703.1
4, 5
1013
XM_017015216.1
XP_016870705.1
4, 5
1013
GenBank mRNA
AB058760.2
-
2, 3
992
AY358165.1
-
3, 4
562
BC013770.1
-
1, 2
301
4344617
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix protein
Gene Ontology Categories:
Function(s)
Process(es)
axonogenesis
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0