Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201435.4 | NP_958843.2 | 10 | 1898 | ||
NR_027918.2 | 10 | 1898 | ||||
XM_006719643.1 | XP_006719706.1 | 10 | 1806 | |||
XM_006719644.1 | XP_006719707.1 | 10 | 1806 | |||
XM_011538856.1 | XP_011537158.1 | 10 | 1803 | |||
XM_011538857.1 | XP_011537159.1 | 10 | 1806 | |||
XM_011538859.1 | XP_011537161.1 | 7 | 1269 | |||
XM_017020033.1 | XP_016875522.1 | 9 | 2684 | |||
GenBank mRNA | AK097663.1 | - | 10 | 1822 | ||
AK124633.1 | - | 6 | 2222 | |||
AK292466.1 | - | 10 | 1806 | |||
AY009105.1 | - | 6 | 1214 | |||
AY254201.1 | - | 10 | 1879 | |||
BC060796.1 | - | 10 | 1881 | 5284906 |