Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127219.1 | NP_001120691.1 | 5, 6 | 634 | ||
NM_032592.3 | NP_115981.1 | 5, 6 | 930 | |||
XM_005253170.3 | XP_005253227.2 | 5, 6 | 1073 | |||
XM_005253172.2 | XP_005253229.1 | 2, 3 | 161 | |||
XM_006718348.2 | XP_006718411.1 | 5, 6 | 426 | |||
XM_011520406.2 | XP_011518708.1 | 5, 6 | 1076 | |||
XM_011520407.1 | XP_011518709.1 | 5, 6 | 426 | |||
XM_011520410.2 | XP_011518712.1 | 5, 6 | 1092 | |||
XM_017018421.1 | XP_016873910.1 | 5, 6 | 1077 | |||
GenBank mRNA | AK057649.1 | - | 5, 6 | 924 | ||
AK302515.1 | - | 5, 6 | 441 | |||
AK304074.1 | - | 4, 5 | 796 | |||
AK307380.1 | - | 5, 6 | 630 | |||
AY026508.1 | - | 5, 6 | 885 | |||
BC020197.1 | - | 5, 6 | 904 | 3632065 | ||
KU178811.1 | - | 4, 5 | 490 | |||
KU178812.1 | - | 4, 5 | 490 | |||
KU178813.1 | - | 3, 4 | 430 |