Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032600.2 | NP_115989.1 | 1 | 368 | ||
GenBank mRNA | AF367469.1 | - | 1 | 264 | ||
AK058091.1 | - | 1 | 368 |