Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168399.1 | NP_001161871.1 | 3 | 486 | ||
NM_001168400.1 | NP_001161872.1 | 3 | 483 | |||
NM_001168401.1 | NP_001161873.1 | 3 | 228 | |||
NM_032621.3 | NP_116010.1 | 3 | 300 | |||
GenBank mRNA | AF251053.1 | - | 3 | 132 | ||
AK312085.1 | - | 3 | 224 | |||
AY833560.1 | - | 3 | 232 | |||
BC015522.1 | - | 3 | 178 | 3914401 | ||
BI670202.1 | - | 3 | 279 | |||
BU569325.1 | - | 3 | 303 | |||
BU596902.1 | - | 3 | 211 | |||
CB961647.1 | - | 3 | 492 | |||
CD365521.1 | - | 1 | 611 | |||
CK001963.1 | - | 3 | 231 |