Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168399.1 | NP_001161871.1 | 3 | 577 | ||
NM_001168400.1 | NP_001161872.1 | 3 | 574 | |||
NM_001168401.1 | NP_001161873.1 | 3 | 319 | |||
NM_032621.3 | NP_116010.1 | 3 | 391 | |||
GenBank mRNA | AF251053.1 | - | 3 | 223 | ||
AK312085.1 | - | 3 | 315 | |||
AY833560.1 | - | 3 | 323 | |||
BC015522.1 | - | 3 | 269 | 3914401 | ||
BI670202.1 | - | 3 | 370 | |||
BU569325.1 | - | 3 | 394 | |||
BU596902.1 | - | 3 | 302 | |||
CB961647.1 | - | 3 | 583 | |||
CD365521.1 | - | 1 | 520 | |||
CK001963.1 | - | 3 | 322 |