Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256421.1 | NP_001243350.1 | 2 | 275 | ||
NM_001256422.1 | NP_001243351.1 | 2 | 279 | |||
NM_032717.4 | NP_116106.2 | 1 | 362 | |||
XM_011532384.2 | XP_011530686.1 | 2 | 274 | |||
XM_017008780.1 | XP_016864269.1 | 2 | 431 | |||
XM_017008781.1 | XP_016864270.1 | 2 | 438 | |||
GenBank mRNA | AK055749.1 | - | 2 | 274 | ||
AK223543.1 | - | 2 | 246 | |||
AY358100.1 | - | 1 | 179 | |||
BC090956.1 | - | 1 | 268 | 30387839 | ||
DA393801.1 | - | 2 | 279 | |||
DA673378.1 | - | 1 | 362 | |||
DQ324782.1 | - | 1 | 59 | |||
DQ345298.1 | - | 1 | 359 |